Kobalt, Sigma-Aldrich®
Dodavatel: Merck
|
Nebezpečí
|
M-Clarity™ quality level = MQ100
266655-100G
266655-500G
SIAL266655-100GEA
2620
CZK
SIAL266655-100G
SIAL266655-500G
Kobalt, Sigma-Aldrich®
Kobalt
Vzorec:
Co MW: 58.93 Bod varu: 2870 Bod tání: 1495 Hustota: 8.9 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00010935 CAS číslo: 7440-48-4 EINECS: 231-158-0 Merck Index: 12,02488 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...