Tisk…
Kobalt ≥99.5% (metals basis), powder -60 mesh
Dodavatel: Thermo Fisher Scientific
Nebezpečí
10456.22 10456.36
10456.22EA 4880 CZK
10456.22 10456.36
Kobalt ≥99.5% (metals basis), powder -60 mesh
Kobalt
Purity: 99.5% CAS No.: 7440-48-4 Appearance: Black Odour: Odourless UN No.: 3089 MDL No.: MFCD00010935
Vzorec: Co
Storage Temperature: Ambient
MDL Number: MFCD00010935
CAS číslo: 7440-48-4
EINECS: 231-158-0
UN: 3089
ADR: 4.1,III