Manganese ≥99.95% (metals basis), powder -325 mesh
Dodavatel: Thermo Fisher Scientific
|
Varování
|
36315.14
36315.22
36315.14EA
4090
CZK
36315.14
36315.22
Manganese ≥99.95% (metals basis), powder -325 mesh
Manganese
Purity: 99.95%
CAS No.: 7439-96-5
UN No.: 3089
MDL No.: MFCD00011111
Vzorec:
Mn Bod varu: 2097 °C (1013 hPa) Bod tání: 1260 °C Hustota: 7,2 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011111 CAS číslo: 7439-96-5 EINECS: 231-105-1 UN: 3089 ADR: 4.1,III Merck Index: 12,05762 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...