Tisk…
Manganese ≥99.98% (metals basis), granules 0.8-12 mm (0.03-0.47 in)
Dodavatel: Thermo Fisher Scientific
36215.14 36215.22
36215.14EA 3640 CZK
36215.14 36215.22
Manganese ≥99.98% (metals basis), granules 0.8-12 mm (0.03-0.47 in)
Manganese
Purity: 99.98% CAS No.: 7439-96-5 Appearance: Grey Odour: Odourless MDL No.: MFCD00011111
For vacuum deposition
Vzorec: Mn
MW: 54.94
Storage Temperature: Ambient
MDL Number: MFCD00011111
CAS číslo: 7439-96-5
EINECS: 231-105-1