Tisk…
Molybden, gauze woven from wire 100 mesh, Ø 0.0636 mm (0.0025 in)
Dodavatel: Thermo Fisher Scientific
40912.FL 40912.GT 40912.HB
40912.FLEA 6830 CZK
40912.FL 40912.GT 40912.HB
Molybden, gauze woven from wire 100 mesh, Ø 0.0636 mm (0.0025 in)
Molybden
CAS No.: 7439-98-7 Physical Form: Wire Cloth MDL No.: MFCD00003465
Vzorec: Mo
MW: 95.94
Bod varu: 3617
Bod tání: 2622
Hustota: 10.28 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003465
CAS číslo: 7439-98-7
EINECS: 231-107-2
Merck Index: 12,06317