Tisk…
Molybden ≥99.95% (metals basis), plate, Thickness: 2.5 mm (0.098 in)
Dodavatel: Thermo Fisher Scientific
10047.FI 10047.GH 10047.HP
10047.FIEA 4530 CZK
10047.FI 10047.GH 10047.HP
Molybden ≥99.95% (metals basis), plate, Thickness: 2.5 mm (0.098 in)
Molybden
Purity: 99.95% CAS No.: 7439-98-7 Physical Form: ≈64g/50x50mm MDL No.: MFCD00003465
Vzorec: Mo
MW: 95.94
Bod varu: 3617
Bod tání: 2622
Hustota: 10.28 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003465
CAS číslo: 7439-98-7
EINECS: 231-107-2
Merck Index: 12,06317