Vanad ≥99.7% (metals basis), pieces
Dodavatel: Thermo Fisher Scientific
42775.09
42775.18
42775.30
42775.09EA
2770
CZK
42775.09
42775.18
42775.30
Vanad ≥99.7% (metals basis), pieces
Vanad
Purity: 99.7%
CAS No.: 7440-62-2
Odour: Odourless
MDL No.: MFCD00011453
Vzorec:
V Bod varu: 3477 °C (1013 hPa) Bod tání: 1896 °C Hustota: 6,015 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011453 CAS číslo: 7440-62-2 EINECS: 231-171-1 Merck Index: 12,10053 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...