Tisk…

Tantalum ≥99.9% (metals basis), powder APS ∼2 µm

Dodavatel: Thermo Fisher Scientific
Nebezpečí

39690.06 39690.14 39690.18
39690.06EA 3260 CZK
39690.06 39690.14 39690.18
Tantalum ≥99.9% (metals basis), powder APS ∼2 µm
Tantalum

Purity: 99.9% CAS No.: 7440-25-7 Appearance: Black Odour: Odourless UN No.: 3089 MDL No.: MFCD00011252

Vzorec: Ta
Hustota: 16,6 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011252
CAS číslo: 7440-25-7
EINECS: 231-135-5
UN: 3089
ADR: 4.1,II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR