Tantalum ≥99.9% (metals basis), wire, hard, Ø 0.203 mm (0.008 in)
Dodavatel: Thermo Fisher Scientific
45133.G1
45133.G6
45133.G1EA
2100
CZK
45133.G1
45133.G6
Tantalum ≥99.9% (metals basis), wire, hard, Ø 0.203 mm (0.008 in)
Tantalum
Purity: 99.9%
CAS No.: 7440-25-7
Molecular Formula: Ta
Formula Weight: 180.95
Physical Form: ca 0.54g/m
Density: 16.69
Melting Point: 2996°
Boiling Point: 5429°
MDL No.: MFCD00011252
Vzorec:
Ta Bod varu: 5429 °C Bod tání: 2996 °C Hustota: 16,69 Storage Temperature: Ambient |
MDL Number:
MFCD00011252 CAS číslo: 7440-25-7 EINECS: 231-135-5 Merck Index: 12,09223 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...