Tisk…

Tantalum ≥99.95% (metals basis), rods, Ø 2.8 mm (0.11 in)

Dodavatel: Thermo Fisher Scientific

42371.BU 42371.BY
42371.BUEA 7350 CZK
42371.BU 42371.BY
Tantalum ≥99.95% (metals basis), rods, Ø 2.8 mm (0.11 in)
Tantalum

Purity: 99.95% CAS No.: 7440-25-7 Physical Form: ≈102g/m Odour: Odourless MDL No.: MFCD00011252

Vzorec: Ta
Bod varu: 5427 °C (1013 hPa)
Bod tání: 2996 °C
Hustota: 16,69 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011252
CAS číslo: 7440-25-7
EINECS: 231-135-5
Merck Index: 12,09223

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR