Tisk…

Tantalum ≥99.9% (metals basis), foil, Thickness: 0.7 mm (0.03 in)

Dodavatel: Thermo Fisher Scientific

00911.FG 00911.FY
00911.FGEA 4880 CZK
00911.FG 00911.FY
Tantalum ≥99.9% (metals basis), foil, Thickness: 0.7 mm (0.03 in)
Tantalum

Purity: 99.9% CAS No.: 7440-25-7 Physical Form: ≈14.5g/25x50mm Odour: Odourless MDL No.: MFCD00011252

Vzorec: Ta
Bod varu: 5427 °C (1013 hPa)
Bod tání: 2996 °C
Hustota: 16,69 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011252
CAS číslo: 7440-25-7
EINECS: 231-135-5
Merck Index: 12,09223

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR