Tisk…
Niob ≥99.8% (metals basis excluding Ta), powder -60 mesh (max. 1% Ta)
Dodavatel: Thermo Fisher Scientific
10276.14 10276.22 10276.36
10276.14EA 2080 CZK
10276.14 10276.22 10276.36
Niob ≥99.8% (metals basis excluding Ta), powder -60 mesh (max. 1% Ta)
Niobium
Purity: 99.8% CAS No.: 7440-03-1 Odour: Odourless MDL No.: MFCD00011126
Vzorec: Nb
MW: 92.91
Bod varu: 3700
Bod tání: 2468
Storage Temperature: Ambient
MDL Number: MFCD00011126
CAS číslo: 7440-03-1
EINECS: 231-113-5
UN: 3089
ADR: 4.1 II