Tisk…
Niob ≥99.8% (metals basis), rods, hard, Ø 12.7 mm (0.5 in)
Dodavatel: Thermo Fisher Scientific
43362.BS 43362.BP
43362.BSEA 11840 CZK
43362.BS 43362.BP
Niob ≥99.8% (metals basis), rods, hard, Ø 12.7 mm (0.5 in)
Niobium
Purity: 99.8% CAS No.: 7440-03-1 Physical Form: ≈10.9g/cm, Temper: as drawn Odour: Odourless MDL No.: MFCD00011126
Vzorec: Nb
MW: 92.91
Bod varu: 3700
Bod tání: 2468
Hustota: 8.57
Storage Temperature: Ambient
MDL Number: MFCD00011126
CAS číslo: 7440-03-1
EINECS: 231-113-5