Tisk…
Niob ≥99.96% (metals basis excluding Ta), wire, Ø 0.5 mm (0.02 in)
Dodavatel: Thermo Fisher Scientific
14335.BU 14335.BY 14335.CC
14335.BUEA 1880 CZK
14335.BU 14335.BY 14335.CC
Niob ≥99.96% (metals basis excluding Ta), wire, Ø 0.5 mm (0.02 in)
Niobium
Purity: 99.96% CAS No.: 7440-03-1 Physical Form: ≈1.68g/m Odour: Odourless MDL No.: MFCD00011126
Vzorec: Nb
MW: 92.91
Bod varu: 3700
Bod tání: 2468
Hustota: 8.57 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011126
CAS číslo: 7440-03-1
EINECS: 231-113-5