Tisk…
Zirconium ≥99.2% (metals basis excluding Hf), wire (max. 4.5% Hf), annealed, Ø 0.5 mm (0.020 in)
Dodavatel: Thermo Fisher Scientific
45056.G5 45056.G1
45056.G5EA 3380 CZK
45056.G5 45056.G1
Zirconium ≥99.2% (metals basis excluding Hf), wire (max. 4.5% Hf), annealed, Ø 0.5 mm (0.020 in)
Zirconium
Purity: 99.2% CAS No.: 7440-67-7 Physical Form: ≈1.31g/m Odour: Odourless MDL No.: MFCD00011303
Vzorec: Zr
MW: 91.22
Storage Temperature: Ambient
MDL Number: MFCD00011303
CAS číslo: 7440-67-7
EINECS: 231-176-9