Tisk…

Zirconium ≥99.9% (metals basis), foil, Thickness: 0.025 mm (0.001 in)

Dodavatel: Thermo Fisher Scientific

43049.FQ 43049.FF 43049.FW
43049.FQEA 6280 CZK
43049.FQ 43049.FF 43049.FW
Zirconium ≥99.9% (metals basis), foil, Thickness: 0.025 mm (0.001 in)
Zirconium

Purity: 99.9+% CAS No.: 7440-67-7 Physical Form: ≈0.1g/25x25mm Odour: Odourless MDL No.: MFCD00011303

Vzorec: Zr
Storage Temperature: Ambient
MDL Number: MFCD00011303
CAS číslo: 7440-67-7
EINECS: 231-176-9

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR