Tisk…
Zirconium ≥99.95% (metals basis excluding Hf), slug (Hf ∼ 3%), Length: 6.35 mm (0.25 in), Ø 3.175 mm (0.125 in)
Dodavatel: Thermo Fisher Scientific
42558.06 42558.14
42558.06EA 2290 CZK
42558.06 42558.14
Zirconium ≥99.95% (metals basis excluding Hf), slug (Hf ∼ 3%), Length: 6.35 mm (0.25 in), Ø 3.175 mm (0.125 in)
Zirconium
Purity: 99.95% CAS No.: 7440-67-7 Physical Form: ≈0.33g/slug Odour: Odourless MDL No.: MFCD00011303
Vzorec: Zr
MW: 91.22
Storage Temperature: Ambient
MDL Number: MFCD00011303
CAS číslo: 7440-67-7
EINECS: 231-176-9