Tisk…
Zirconium ≥99.95% (metals basis excluding Hf), slug (Hf ∼ 3%), Length: 12.7 mm (0.50 in), Ø 6.35 mm (0.25 in)
Dodavatel: Thermo Fisher Scientific
42556.QJ 42556.QK
42556.QJEA 2810 CZK
42556.QJ 42556.QK
Zirconium ≥99.95% (metals basis excluding Hf), slug (Hf ∼ 3%), Length: 12.7 mm (0.50 in), Ø 6.35 mm (0.25 in)
Zirconium
Purity: 99.95% CAS No.: 7440-67-7 Physical Form: ≈2.62g/slug Odour: Odourless MDL No.: MFCD00011303
Vzorec: Zr
MW: 91.22
Storage Temperature: Ambient
MDL Number: MFCD00011303
CAS číslo: 7440-67-7
EINECS: 231-176-9