Tisk…
Zirconium ≥99.5% (metals basis), foil, annealed, Thickness: 0.127 mm (0.005 in)
Dodavatel: Thermo Fisher Scientific
10594.GM 10594.GR 10594.IB
10594.GMEA 3060 CZK
10594.GM 10594.GR 10594.IB
Zirconium ≥99.5% (metals basis), foil, annealed, Thickness: 0.127 mm (0.005 in)
Zirconium
Purity: 99.5% CAS No.: 7440-67-7 Physical Form: ≈10.3g/100x125mm, Temper: soft Odour: Odourless MDL No.: MFCD00011303
Vzorec: Zr
MW: 91.22
Storage Temperature: Ambient
MDL Number: MFCD00011303
CAS číslo: 7440-67-7
EINECS: 231-176-9