Tisk…
Zirconium ≥99% (metals basis excluding Hf), rods (max. 4.5% Hf), annealed, Ø 6.2 mm (0.24 in)
Dodavatel: Thermo Fisher Scientific
10443.BS 10443.BW
10443.BSEA 1850 CZK
10443.BS 10443.BW
Zirconium ≥99% (metals basis excluding Hf), rods (max. 4.5% Hf), annealed, Ø 6.2 mm (0.24 in)
Zirconium
Purity: 99% CAS No.: 7440-67-7 Physical Form: ≈1.96g/cm, Temper: soft Odour: Odourless MDL No.: MFCD00011303
Vzorec: Zr
MW: 91.22
Storage Temperature: Ambient
MDL Number: MFCD00011303
CAS číslo: 7440-67-7
EINECS: 231-176-9