Tisk…
Zirconium ≥99.2% (metals basis excluding Hf), wire (max. 4.5% Hf), Ø 1.0 mm (0.04 in)
Dodavatel: Thermo Scientific
00939.G2 00939.G6
00939.G2EA 1820 CZK
00939.G2 00939.G6
Zirconium ≥99.2% (metals basis excluding Hf), wire (max. 4.5% Hf), Ø 1.0 mm (0.04 in)
Zirconium
Purity: 99.2% CAS No.: 7440-67-7 Physical Form: ≈5.11g/m Odour: Odourless MDL No.: MFCD00011303
Vzorec: Zr
MW: 91,22 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011303
CAS číslo: 7440-67-7
EINECS: 231-176-9