Tisk…
Zirconium ≥99.2% (metals basis excluding Hf), plate 25x100 mm (0.98x3.9 in), Thickness: 6.4 mm (0.25 in)
Dodavatel: Thermo Fisher Scientific
00934.KF 00934.KJ
00934.KFEA 4500 CZK
00934.KF 00934.KJ
Zirconium ≥99.2% (metals basis excluding Hf), plate 25x100 mm (0.98x3.9 in), Thickness: 6.4 mm (0.25 in)
Zirconium
Purity: 99.2% CAS No.: 7440-67-7 Physical Form: ≈104g/25x100mm, Temper: annealed Odour: Odourless MDL No.: MFCD00011303
Vzorec: Zr
MW: 91.22
Storage Temperature: Ambient
MDL Number: MFCD00011303
CAS číslo: 7440-67-7
EINECS: 231-176-9