Tisk…
Hafnium ≥99.5% (metals basis excluding Zr), foil (Zr ∼ 3%), annealed, Thickness: 0.25 mm (0.01 in)
Dodavatel: Thermo Fisher Scientific
10793.FJ 10793.FP
10793.FJEA 8510 CZK
10793.FJ 10793.FP
Hafnium ≥99.5% (metals basis excluding Zr), foil (Zr ∼ 3%), annealed, Thickness: 0.25 mm (0.01 in)
Hafnium
Purity: 99.5% CAS No.: 7440-58-6 Physical Form: ≈13.3g/50x80mm Odour: Odourless MDL No.: MFCD00011032
Vzorec: Hf
MW: 178.49
Storage Temperature: Ambient
MDL Number: MFCD00011032
CAS číslo: 7440-58-6
EINECS: 231-166-4