Tisk…

Kadmium ≥99.998% (metals basis), wire, Puratronic®, Ø 2.0 mm (0.08 in)

Dodavatel: Thermo Fisher Scientific
Nebezpečí

11908.BU 11908.G1
11908.BUEA 3170 CZK
11908.BU 11908.G1
Kadmium ≥99.998% (metals basis), wire, Puratronic®, Ø 2.0 mm (0.08 in)
Kadmium

Purity: 99.998% CAS No.: 7440-43-9 Physical Form: ≈27.2g/m MDL No.: MFCD00010914

Vzorec: Cd
Bod varu: 765 °C (1013 hPa)
Bod tání: 321 °C
Hustota: 8,642 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010914
CAS číslo: 7440-43-9
EINECS: 231-152-8
UN: 2570
ADR: 6.1,II
Merck Index: 12,01649

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR