Tisk…
Kadmium ≥99.998% (metals basis), foil, Puratronic®, Thickness: 1.0 mm (0.04 in)
Dodavatel: Thermo Fisher Scientific
Nebezpečí
11368.FI 11368.GH
11368.FIEA 6940 CZK
11368.FI 11368.GH
Kadmium ≥99.998% (metals basis), foil, Puratronic®, Thickness: 1.0 mm (0.04 in)
Kadmium
Purity: 99.998% CAS No.: 7440-43-9 Physical Form: ≈21.6g/50x50mm MDL No.: MFCD00010914
Vzorec: Cd
MW: 112.41
Bod varu: 765
Bod tání: 321
Hustota: 8.642 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010914
CAS číslo: 7440-43-9
EINECS: 231-152-8
UN: 2570
ADR: 6.1 II
Merck Index: 12,01649