Tisk…
Kadmium ≥99.999% (metals basis), foil, Puratronic®, Thickness: 0.025 mm (0.001 in)
Dodavatel: Thermo Fisher Scientific
Nebezpečí
00064.FQ 00064.FV
00064.FQEA 3640 CZK
00064.FQ 00064.FV
Kadmium ≥99.999% (metals basis), foil, Puratronic®, Thickness: 0.025 mm (0.001 in)
Kadmium
Purity: 99.999% CAS No.: 7440-43-9 Physical Form: ≈0.54g/25x100mm MDL No.: MFCD00010914
Vzorec: Cd
MW: 112.41
Bod varu: 765
Bod tání: 321
Hustota: 8.642 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010914
CAS číslo: 7440-43-9
EINECS: 231-152-8
UN: 2570
ADR: 6.1 II
Merck Index: 12,01649