stříbro ≥99.998% (metals basis), wire, Premion®, Ø 1.5 mm (0.06 in)
Dodavatel: Thermo Fisher Scientific
42487.BU
42487.BY
42487.CC
42487.BUEA
2990
CZK
42487.BU
42487.BY
42487.CC
stříbro ≥99.998% (metals basis), wire, Premion®, Ø 1.5 mm (0.06 in)
stříbro
Purity: 99.998%
CAS No.: 7440-22-4
Physical Form: ≈18.54g/m
Odour: Characteristic
MDL No.: MFCD00003397
Vzorec:
Ag MW: 107.87 Bod varu: 2212 Bod tání: 962 Hustota: 10.5 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00003397 CAS číslo: 7440-22-4 EINECS: 231-131-3 Merck Index: 12,08647 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...