Tisk…

stříbro ≥99.9% (metals basis), foil, annealed, Thickness: 0.28 mm (0.011 in)

Dodavatel: Thermo Fisher Scientific

41451.FI 41451.GH 41451.GW
41451.FIEA 2540 CZK
41451.FI 41451.GH 41451.GW
stříbro ≥99.9% (metals basis), foil, annealed, Thickness: 0.28 mm (0.011 in)
stříbro

Purity: 99.9% CAS No.: 7440-22-4 Physical Form: ≈7.3g/50x50mm, Temper: soft Odour: Characteristic MDL No.: MFCD00003397

Vzorec: Ag
Bod varu: 2212 °C (1013 hPa)
Bod tání: 962 °C
Hustota: 10,5 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003397
CAS číslo: 7440-22-4
EINECS: 231-131-3
Merck Index: 12,08647

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR