Tisk…

stříbro ≥99.9% (metals basis), powder APS 1-3 µm

Dodavatel: Thermo Fisher Scientific

11405.09 11405.18 11405.30
11405.09EA 2550 CZK
11405.09 11405.18 11405.30
stříbro ≥99.9% (metals basis), powder APS 1-3 µm
stříbro

Purity: 99.9% CAS No.: 7440-22-4 Physical Form: S.A. min 0.25m2/g Odour: Characteristic UN No.: 3077 MDL No.: MFCD00003397

Vzorec: Ag
Bod varu: 2212 °C (1013 hPa)
Hustota: 10,5 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003397
CAS číslo: 7440-22-4
EINECS: 231-131-3
UN: 3077
ADR: 9,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR