stříbro ≥99.999% (metals basis), powder, Premion® -120 mesh
Dodavatel: Thermo Fisher Scientific
00781.06
00781.14
00781.06EA
2960
CZK
00781.06
00781.14
stříbro ≥99.999% (metals basis), powder, Premion® -120 mesh
stříbro
Purity: 99.999%
CAS No.: 7440-22-4
Odour: Characteristic
UN No.: 3077
MDL No.: MFCD00003397
Vzorec:
Ag Bod varu: 2212 °C (1013 hPa) Hustota: 10,5 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00003397 CAS číslo: 7440-22-4 EINECS: 231-131-3 UN: 3077 ADR: 9,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...