Zlato ≥99.95% (metals basis), wire, Ø 1.0 mm (0.04 in)
Dodavatel: Thermo Fisher Scientific
14731.BP
14731.BS
14731.BW
14731.BPEA
2980
CZK
14731.BP
14731.BS
14731.BW
Zlato ≥99.95% (metals basis), wire, Ø 1.0 mm (0.04 in)
Zlato
Purity: 99.95%
CAS No.: 7440-57-5
Physical Form: ≈15.15g/m
Odour: Odourless
MDL No.: MFCD00003436
Vzorec:
Au MW: 196.97 Bod varu: 2800 Bod tání: 1063 Hustota: 19.3 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00003436 CAS číslo: 7440-57-5 EINECS: 231-165-9 Merck Index: 12,04526 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...