Tisk…

Zlato 99,95 Metals basis, wire, Ø 2.0 mm (0.08 in)

Dodavatel: Thermo Fisher Scientific

14729.BO 14729.BQ 14729.BU
14729.BOEA 5980 CZK
14729.BO 14729.BQ 14729.BU
Zlato 99,95 Metals basis, wire, Ø 2.0 mm (0.08 in)
Zlato

Purity: 99.95% CAS No.: 7440-57-5 Physical Form: ≈60.6g/m Odour: Odourless MDL No.: MFCD00003436

Vzorec: Au
MW: 196.97
Bod varu: 2800
Bod tání: 1063
Hustota: 19.3 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003436
CAS číslo: 7440-57-5
EINECS: 231-165-9
Merck Index: 12,04526

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR