Tisk…
ESD safety shoes, lace-up, Uni6, 31721
Dodavatel: Abeba
31721/35 31721/36 31721/37 31721/38 31721/39 31721/40 31721/41 31721/42 31721/43 31721/44 31721/45 31721/46 31721/47 31721/48
ABEB31721/35EA 3740 CZK
ABEB31721/35 ABEB31721/36 ABEB31721/37 ABEB31721/38 ABEB31721/39 ABEB31721/40 ABEB31721/41 ABEB31721/42 ABEB31721/43 ABEB31721/44 ABEB31721/45 ABEB31721/46 ABEB31721/47 ABEB31721/48
ESD safety shoes, lace-up, Uni6, 31721
Obuv
ESD shoes made from velour upper and breathable textile inlays with reflective material and a TPU sole resistant to oil, fuel and animal fats.
  • Perspiration-absorbent special lining with Silver Point
  • Replaceable acc Wave insole; orthopaedic insoles possible
  • Sanitized® treatment reduces odour and bacterial growth
  • Slip resistant TPU sole
  • Protective steel toecap (200 J)
Colour: Black
Certifikace: EN 61340 (ESD); DGUV 112-191; EN ISO 20345:2011; S1, SRC