Tisk…

Ytterbium ≥99.9% (REO, rare earth oxide basis), foil, Thickness: 1.0 mm (0.04 in)

Dodavatel: Thermo Scientific

10426.FF 10426.FI
10426.FFEA 7250 CZK
10426.FF 10426.FI
Ytterbium ≥99.9% (REO, rare earth oxide basis), foil, Thickness: 1.0 mm (0.04 in)
Ytterbium

Purity: 99.9% CAS No.: 7440-64-4 Physical Form: ≈4.4g/25x25mm, Packaged in mylar under argon Odour: Odourless MDL No.: MFCD00011286

Vzorec: Yb
MW: 173,04 g/mol
Bod varu: 1427 °C (1013 hPa)
Bod tání: 824 °C
Hustota: 6,977 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011286
CAS číslo: 7440-64-4
EINECS: 231-173-2
Merck Index: 12,10239

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR