Tisk…

Ytterbium ≥99.9% (REO, rare earth oxide basis), foil, Thickness: 0.64 mm (0.025 in)

Dodavatel: Thermo Fisher Scientific

00614.FF 00614.FI
00614.FFEA 6080 CZK
00614.FF 00614.FI
Ytterbium ≥99.9% (REO, rare earth oxide basis), foil, Thickness: 0.64 mm (0.025 in)
Ytterbium

Purity: 99.9% CAS No.: 7440-64-4 Physical Form: ≈2.8g/25x25mm, Packaged in mylar under argon MDL No.: MFCD00011286

Vzorec: Yb
Bod varu: 1427 °C (1013 hPa)
Bod tání: 824 °C
Hustota: 6,977 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011286
CAS číslo: 7440-64-4
EINECS: 231-173-2
Merck Index: 12,10239

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR