Tisk…

Ytterbium ≥99.9% (REO, rare earth oxide basis), ingot

Dodavatel: Thermo Fisher Scientific

00407.06 00407.14
00407.06EA 3780 CZK
00407.06 00407.14
Ytterbium ≥99.9% (REO, rare earth oxide basis), ingot
Ytterbium

Purity: 99.9% CAS No.: 7440-64-4 Physical Form: Packaged in air stable poly bottle Odour: Odourless MDL No.: MFCD00011286

Vzorec: Yb
MW: 173.04
Bod varu: 1427
Bod tání: 824
Hustota: 6.977 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011286
CAS číslo: 7440-64-4
EINECS: 231-173-2
Merck Index: 12,10239

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR