Tisk…
Thulium ≥99.9% (REO, rare earth oxide basis), foil, Thickness: 0.25 mm (0.01 in)
Dodavatel: Thermo Fisher Scientific
Nebezpečí
Packaged under argon.
10375.FF 10375.FI
10375.FFEA 11190 CZK
10375.FF 10375.FI
Thulium ≥99.9% (REO, rare earth oxide basis), foil, Thickness: 0.25 mm (0.01 in)
Thulium
Purity: 99.9% CAS No.: 7440-30-4 Physical Form: ≈1.75g/25x25mm, Packaged under argon Appearance: Silver-grey Odour: Odourless UN No.: 3178 MDL No.: MFCD00011281
Vzorec: Tm
MW: 168.93
Storage Temperature: Ambient
MDL Number: MFCD00011281
CAS číslo: 7440-30-4
EINECS: 231-140-2
UN: 3178
ADR: 4.1 II