Terbium ≥99.9% (REO, rare earth oxide basis), ingot (max. 0,4% Ta)
Dodavatel: Thermo Fisher Scientific
00320.04
00320.09
00320.04EA
3850
CZK
00320.04
00320.09
Terbium ≥99.9% (REO, rare earth oxide basis), ingot (max. 0,4% Ta)
Terbium
Purity: 99.9%
CAS No.: 7440-27-9
Physical Form: Packaged in air-stable poly bottle
MDL No.: MFCD00011256
Vzorec:
Tb Bod varu: 3041 °C (1013 hPa) Bod tání: 1356 °C Hustota: 8,27 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011256 CAS číslo: 7440-27-9 EINECS: 231-137-6 Merck Index: 12,09300 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...