Samárium, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ100
261203-10G
261203-1G
261203-50G
SIAL261203-10GEA
12520
CZK
SIAL261203-10G
SIAL261203-1G
SIAL261203-50G
Samárium, Sigma-Aldrich®
Samárium
Vzorec:
Sm Bod varu: 1900 °C (1013 hPa) Hustota: 7,42 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011233 CAS číslo: 7440-19-9 EINECS: 231-128-7 Merck Index: 12,08498 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...