Samárium ≥99.9% (REO, rare earth oxide basis), foil, Thickness: 1.0 mm (0.04 in)
Dodavatel: Thermo Fisher Scientific
43217.FF
43217.FI
43217.FFEA
12080
CZK
43217.FF
43217.FI
Samárium ≥99.9% (REO, rare earth oxide basis), foil, Thickness: 1.0 mm (0.04 in)
Samárium
Purity: 99.9%
CAS No.: 7440-19-9
Physical Form: ≈4.9g/25x25mm, Packaged in mylar under argon
Odour: Odourless
MDL No.: MFCD00011233
Vzorec:
Sm MW: 150.36 Bod varu: 1900 Bod tání: 1076.85 Hustota: 7.42 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011233 CAS číslo: 7440-19-9 EINECS: 231-128-7 Merck Index: 12,08498 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...