Samárium ≥99.9% (REO, rare earth oxide basis), pieces ≤ 12 mm (0.47 in)
Dodavatel: Thermo Fisher Scientific
00609.09
00609.18
00609.09EA
3350
CZK
00609.09
00609.18
Samárium ≥99.9% (REO, rare earth oxide basis), pieces ≤ 12 mm (0.47 in)
Samárium
Purity: 99.9%
CAS No.: 7440-19-9
Physical Form: Packaged in air stable poly bottle
Sensitivity: Air and Moisture Sensitive
Appearance: Grey
Odour: Odourless
MDL No.: MFCD00011233
Vzorec:
Sm Bod varu: 1900 °C (1013 hPa) Hustota: 7,42 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011233 CAS číslo: 7440-19-9 EINECS: 231-128-7 Merck Index: 12,08498 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...