Tisk…

Samárium ≥99.9% (REO, rare earth oxide basis), ingot

Dodavatel: Thermo Fisher Scientific

00295.14 00295.22
00295.14EA 4840 CZK
00295.14 00295.22
Samárium ≥99.9% (REO, rare earth oxide basis), ingot
Samárium

Purity: 99.9% CAS No.: 7440-19-9 Physical Form: Packaged in air stable poly bottle Odour: Odourless MDL No.: MFCD00011233

Vzorec: Sm
Bod varu: 1900 °C (1013 hPa)
Hustota: 7,42 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011233
CAS číslo: 7440-19-9
EINECS: 231-128-7
Merck Index: 12,08498

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR