Tisk…
Neodymium ≥99.5% (metals basis excluding Ta), rods, Ø 12.7 mm (0.5 in)
Dodavatel: Thermo Fisher Scientific
40294.DH 40294.DJ 40294.DM 40294.HF
40294.DHEA 8210 CZK
40294.DH 40294.DJ 40294.DM 40294.HF
Neodymium ≥99.5% (metals basis excluding Ta), rods, Ø 12.7 mm (0.5 in)
Neodymium
Purity: 99.5% CAS No.: 7440-00-8 Molecular Formula: Packaged in mylar under argon Physical Form: ≈0.89g/mm, Cast rod, diameter is approximate Odour: Odourless
Vzorec: Nd
MW: 144.24
Storage Temperature: Ambient
MDL Number: MFCD00011130
CAS číslo: 7440-00-8
EINECS: 231-109-3