Tisk…

Neodymium ≥99.9% (REO, rare earth oxide basis), foil, Thickness: 0.3 mm (0.012 in)

Dodavatel: Thermo Fisher Scientific

10247.FF 10247.FI
10247.FFEA 8410 CZK
10247.FF 10247.FI
Neodymium ≥99.9% (REO, rare earth oxide basis), foil, Thickness: 0.3 mm (0.012 in)
Neodymium

Purity: 99.9% CAS No.: 7440-00-8 Physical Form: ≈1.3g/25x25mm, Packaged in mylar under argon Odour: Odourless MDL No.: MFCD00011130

Vzorec: Nd
Storage Temperature: Ambient
MDL Number: MFCD00011130
CAS číslo: 7440-00-8
EINECS: 231-109-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR