Tisk…

Lutetium ≥99.9% (REO, rare earth oxide basis), pieces, distilled dendritic

Dodavatel: Thermo Fisher Scientific
Nebezpečí


Packaged under argon.

12410.03 12410.06
12410.03EA 6700 CZK
12410.03 12410.06
Lutetium ≥99.9% (REO, rare earth oxide basis), pieces, distilled dendritic
Lutetium

Purity: 99.9% CAS No.: 7439-94-3 Physical Form: Packaged under argon Appearance: Silver Odour: Odourless UN No.: 3178 MDL No.: MFCD00011098

Vzorec: Lu
Bod varu: 3000 °C (1013 hPa)
Bod tání: 1651…1675 °C
Hustota: 9,849 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011098
CAS číslo: 7439-94-3
EINECS: 231-103-0
UN: 3178
ADR: 4.1,II
Merck Index: 12,05642

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR