Tisk…

Lutetium ≥99.9% (REO, rare earth oxide basis), pieces

Dodavatel: Thermo Fisher Scientific

00628.03 00628.06
00628.03EA 8410 CZK
00628.03 00628.06
Lutetium ≥99.9% (REO, rare earth oxide basis), pieces
Lutetium

Purity: 99.9% CAS No.: 7439-94-3 Physical Form: Packaged in air-stable glass bottle Odour: Odourless MDL No.: MFCD00011098

Vzorec: Lu
Bod varu: 3000 °C (1013 hPa)
Bod tání: 1651…1675 °C
Hustota: 9,849 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011098
CAS číslo: 7439-94-3
EINECS: 231-103-0
Merck Index: 12,05642

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR