Tisk…

Holmium ≥99.9% (REO, rare earth oxide basis), pieces, distilled dendritic

Dodavatel: Thermo Fisher Scientific
Nebezpečí


Packaged under argon.

41515.04 41515.09
41515.04EA 2500 CZK
41515.04 41515.09
Holmium ≥99.9% (REO, rare earth oxide basis), pieces, distilled dendritic
Holmium

Purity: 99.9% CAS No.: 7440-60-0 Physical Form: Packaged under argon Appearance: Dark gray to silver Odour: Odourless UN No.: 3178 MDL No.: MFCD00011049

Vzorec: Ho
Storage Temperature: Ambient
MDL Number: MFCD00011049
CAS číslo: 7440-60-0
EINECS: 231-169-0
UN: 3178
ADR: 4.1,II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR