Tisk…

Gadolinium ≥99.9% (REO, rare earth oxide basis), foil, Thickness: 0.075 mm (0.003 in)

Dodavatel: Thermo Fisher Scientific
Nebezpečí


Packaged under argon.

43307.FF 43307.FI
43307.FFEA 4470 CZK
43307.FF 43307.FI
Gadolinium ≥99.9% (REO, rare earth oxide basis), foil, Thickness: 0.075 mm (0.003 in)
Gadolinium

Purity: 99.9% CAS No.: 7440-54-2 Physical Form: ≈0.36g/25x25mm, Packaged under argon Appearance: Gray UN No.: 3178 MDL No.: MFCD00011022

Vzorec: Gd
MW: 157.25
Storage Temperature: Ambient
MDL Number: MFCD00011022
CAS číslo: 7440-54-2
EINECS: 231-162-2
UN: 3178
ADR: 4.1 II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR