Gadolinium ≥99.9% (REO, rare earth oxide basis), pieces ≤ 20 mm (0.79 in)
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
Packaged under argon.
00118.09
00118.18
00118.09EA
4230
CZK
00118.09
00118.18
Gadolinium ≥99.9% (REO, rare earth oxide basis), pieces ≤ 20 mm (0.79 in)
Gadolinium
Purity: 99.9%
CAS No.: 7440-54-2
Physical Form: Packaged under argon in glass bottle
Appearance: Silver
UN No.: 3178
MDL No.: MFCD00011022
Vzorec:
Gd Storage Temperature: Ambient |
MDL Number:
MFCD00011022 CAS číslo: 7440-54-2 EINECS: 231-162-2 UN: 3178 ADR: 4.1,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...