Tisk…

Europium ≥99.9% (metals basis), rods, Ø 9.52 mm (0.375 in)

Dodavatel: Thermo Fisher Scientific
Nebezpečí

44545.HF 44545.DJ
44545.HFEA 14310 CZK
44545.HF 44545.DJ
Europium ≥99.9% (metals basis), rods, Ø 9.52 mm (0.375 in)
Europium

Purity: 99.9% CAS No.: 7440-53-1 Molecular Formula: Packaged in mineral oil Physical Form: ≈0.37g/mm, Cast rod, diameter approximate Appearance: Silver/gray Odour: Odourless UN No.: 3208 MDL No.: MFCD00010992

Vzorec: Eu
Storage Temperature: Ambient
MDL Number: MFCD00010992
CAS číslo: 7440-53-1
EINECS: 231-161-7
UN: 3208
ADR: 4.3,II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR